Learn More
(±)-U-50488 hydrochloride, Tocris Bioscience™
Selective κ agonist
Brand:  Tocris (Bio-Techne) 049525
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 25mg
 View more versions of this product
Product Code. 13427733
  • £148.47 / 25mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Selective κ-opioid agonist. At higher concentrations blocks Na+ channels.
(-)-enantiomer and (+)-enantiomer also available.
Specifications
Chemical Identifiers
67197-96-0
C19H26Cl2N2O·HCl
+/--u-50488 hydrochloride, +-u-50488 hydrochloride, +-trans-1r,2r-u-50488 hydrochloride, u-50488 hydrochloride, u50488 hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1r,2r-2-pyrrolidin-1-yl cyclohexyl acetamide hydrochloride, c19h26cl2n2o.hcl, +-trans-1r,2r0-u-50488 hcl, trans-2-3,4-dichlorophenyl-n-methyl-n-2-pyrrolidin-1-yl cyclohexyl acetamide hcl
Specifications
25mg
67197-96-0
+/--u-50488 hydrochloride, +-u-50488 hydrochloride, +-trans-1r,2r-u-50488 hydrochloride, u-50488 hydrochloride, u50488 hydrochloride, 2-3,4-dichlorophenyl-n-methyl-n-1r,2r-2-pyrrolidin-1-yl cyclohexyl acetamide hydrochloride, c19h26cl2n2o.hcl, +-trans-1r,2r0-u-50488 hcl, trans-2-3,4-dichlorophenyl-n-methyl-n-2-pyrrolidin-1-yl cyclohexyl acetamide hcl
>99%
(±)-U-50488 hydrochloride
C19H26Cl2N2O·HCl
405.79