Learn More
DuPont™ Tyvek 500 Laboratory Coats press studs
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Collar provide an excellent barrier against fine particles and fibres, is ultra-low-linting and antistatically treated.
Brand: DuPont™ D13396050
Additional Details : Weight : 0.01000kg
Description
- Silicon non-added
- Stitched internal seams
- 3 pockets and 5 press stud closures
- Composed of flash spun high density polyethylene
- Provides an idea balance of protection, durability and comfort
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
X-Large | |
White | |
Single-use | |
3 | |
Stitched (internal) | |
PPE Category, Antistatic treatment (EN 1149-5) - on both sides |
HDPE | |
Press Studs | |
Chemicals | |
Shirt collar | |
Straight cut | |
Pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up. |