Learn More
DuPont™ Tyvek 500 Laboratory Coats press studs
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Collar provide an excellent barrier against fine particles and fibres, is ultra-low-linting and antistatically treated.
Brand: DuPont™ D13396079
235.50 GBP valid until 2024-03-30
Use promo code "21722" to get your promotional price.
Code : NEW
Additional Details : Weight : 5.80000kg
Description
- Silicon non-added
- Stitched internal seams
- 3 pockets and 5 press stud closures
- Composed of flash spun high density polyethylene
- Provides an idea balance of protection, durability and comfort
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Medium |