Learn More
DuPont™ Tyvek™ 400 Dual Hooded Coverall
Provides an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 400 Dual Hooded Coverall is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size.
Brand:  DuPont™ D14809637
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15378354
  • £554.00 / Pack of 100
Estimated Shipment
23-07-2018
Add to basket
Description
Description
  • Coverall with 3-piece hood
  • Tyvek™ at the front and large breathable SMS back
  • Stitched external seams
  • Elasticated wrists, ankles and face
  • Elasticated waist (stitched-in)
  • Tyvek™ zipper and flap
  • White
  • Antistatic
Specifications
Specifications
(EU) 2016/425, EN 1149-5
Tyvek™ fabric front, SMS back
Elastic
Elastic
White
Elastic
Category III, Type 5 and 6
Zipper and flap
108 to 116cm
Hooded
Stitched
X-Large
Elastic
Hooded Coverall
Videos