missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tyrphostin C15, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 mg
Description
A selective ErbB2 inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 149092-50-2 |
Color | Yellow to Orange |
Molecular Formula | C19H15N3O3S2 |
MDL Number | MFCD01074971 |
Quantity | 10 mg |
Sensitivity | Light sensitive |
Synonym | tyrphostin ag 825, tyrphostin c15, 4-hydroxy-3-methoxy-5-benzothiazolylthiomethyl benzylidenecyanoacetamide, 5-benzothiazol-2-yl thiomethyl-4-hydroxy-3-methoxybenzylidenecyanoacetamide, 2e-3-3-1,3-benzothiazol-2-ylsulfanyl methyl-4-hydroxy-5-methoxyphenyl-2-cyanoprop-2-enamide, tocris-1555, biomolki_000008, biomolki2_000018, bmk1-b8, tyrphostin ag |
InChI Key | KXDONFLNGBQLTN-UHFFFAOYSA-N |
SMILES | COC1=CC(C=C(C#N)C(N)=O)=CC(CSC2=NC3=CC=CC=C3S2)=C1O |
IUPAC Name | 3-{3-[(1,3-benzothiazol-2-ylsulfanyl)methyl]-4-hydroxy-5-methoxyphenyl}-2-cyanoprop-2-enamide |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'