Learn More
Tyrphostin B44, (-) enantiomer, Tocris Bioscience™
EGFR-kinase inhibitor
Brand:  Tocris (Bio-Techne) 057810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487773
  • £114.00 / 10mg
Estimated Shipment
28-06-2018
Add to basket
Description
Description
Potent inhibitor of epidermal growth factor receptor (EGFR) kinase (IC50 = 0.4 μM), more active than the (+) enantiomer (Cat. No. 0579). Selective over ErbB2.
Specifications
Chemical Identifiers
Tyrphostin B44, (-) enantiomer
C18H16N2O3
308.34
(R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Specifications
Tyrphostin B44, (-) enantiomer
>98%
308.34
10mg
133550-32-0
C18H16N2O3
(R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
Videos