missing translation for 'onlineSavingsMsg'
Learn More

Tyramine, 98+%, Thermo Scientific Chemicals

Catalog Number 15403669
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
51-67-2
C8H11NO
137.182
MFCD00008193
DZGWFCGJZKJUFP-UHFFFAOYSA-N
tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine
5610
CHEBI:15760
4-(2-aminoethyl)phenol
C1=CC(=CC=C1CCN)O
This item is not returnable. View return policy
51-67-2
C8H11NO
137.182
MFCD00008193
DZGWFCGJZKJUFP-UHFFFAOYSA-N
tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine
5610
CHEBI:15760
4-(2-aminoethyl)phenol
C1=CC(=CC=C1CCN)O

Reagent for fluorescence enzyme immunoassay

Tyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyramine acts as a adrenergic and It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products. It Is a vaso active aminoacid found in foods and it It precipitates headaches in some sufferers.

Solubility
Soluble in water (10 mg/ml at 15°C), methanol, DMSO, boiling alcohol (100 mg/ml), and ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 162°C
Color White to Yellow
Boiling Point 175°C to 181°C (8 mmHg) Hg
Quantity 1 g
Beilstein 1099914
Merck Index 14,9835
Solubility Information Soluble in water (10mg/ml at 15°C),methanol,DMSO,boiling alcohol (100mg/ml),and ethanol.
Formula Weight 137.18
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Tyramine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'