missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Type I 18/10 Stainless Steel Beaker
Manufactured from 18/10 stainless steel. Bochem™ Type I 18/10 Stainless Steel Beaker include a handle and spout.
Brand: Bochem™ 8652
This item is not returnable.
View return policy
Description
- Manufactured from 18/10 stainless steel
- Conical shape
- With handle and spout
Specifications
170 mm | |
Stainless Steel (18/10) | |
Handle, Spout | |
Yes |
2000 mL | |
140 mm | |
Beaker | |
Conical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction