missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Type I 18/10 Stainless Steel Beaker

Manufactured from 18/10 stainless steel. Bochem™ Type I 18/10 Stainless Steel Beaker include a handle and spout.

Brand:  Bochem™ 8651

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10070921

  • £36.42 / Each


Estimated Shipment
22-12-2020
Add to basket

Description

Description

  • Manufactured from 18/10 stainless steel
  • Conical shape
  • With handle and spout
Specifications

Specifications

Conical
1000 mL
125 mm
Beaker
140 mm
Stainless Steel (18/10)
Handle, Spout
Videos
SDS
Documents