missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ Type 90 Safety Storage Underbench Cabinets

Safely store flammable liquids under workbenches, worktops, counters or fume cupboards in accordance with EN 14470-1. With a height of 800mm these cabinets have maximized interior heights making them ideal for storage of larger containers. 

Brand:  Asecos™ 32658-001-30474

Code : NEW

Additional Details : Weight : 115.90000kg

 View more versions of this product

Product Code. 15345579

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

EN 14470-1 with a fire resistance of 90 minutes
482 x 470 x 702mm
800mm
One pullout drawer and perforated metal plate insert
143kg
470mm
Steel
482mm
Storage of flammable liquids
702mm
Powder-coated
593mm
Light Gray
Videos