missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet V-MOVE-90 Light Gray
Safe and approved storage of flammable hazardous substances in working areas
£4280.81 - £5998.54
Specifications
Depth (Metric) Exterior | 860 mm |
---|---|
Depth (Metric) Interior | 775 mm |
Description | Light gray vertical drawer doors |
Color | Light Gray |
Material | Steel |
Alert:
For indoor use only
Product Code | Brand | No. of Doors | Configuration | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Doors | Configuration | Price | Quantity & Availability | ||||
15977726
|
asecos™
3503400135100 |
1 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) |
£4288.23
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15997726
|
asecos™
3503400136111 |
1 | 4 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) |
£4280.81
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15987726
|
asecos™
3503400135101 |
1 | 5 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated) |
£4329.04
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15907736
|
asecos™
3503400136112 |
1 | 5 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated) |
£4329.57
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15937736
|
asecos™
3503500135102 |
2 | 4 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), on each side |
£5915.86
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15957736
|
asecos™
3503500136113 |
2 | 4 x tray shelf, 1 x bottom collecting sump (sheet steel powder-coated), on each side |
£5901.02
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15977736
|
asecos™
3503500136115 |
2 | 5 x tray shelf, 1 x bottom collecting sump, (sheet steel powder-coated), on each side |
£5998.54
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawer opens and closes at the push of a button
- Perfect access: the vertical drawer provides an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawer lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
860 mm | |
Light gray vertical drawer doors | |
Steel | |
Auto-close, Sliding | |
1966 mm | |
Powder-coated plain |
775 mm | |
Light Gray | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1750 mm |