missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 Right-Hinged

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3024000134414

Additional Details : Weight : 207.86850kg

 View more versions of this product

Product Code. 15983906

  • £2055.34 / Each
Estimated Shipment: 03-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

For indoor use only


Description

Description

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

1072 mm
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
524 mm
Steel
616 x 596 x 1298 mm
524 x 450 x 1072 mm
Standard
1298 mm
S90.129.060.R
1
Medium
Safety storage cabinet
450 mm
Powder-coated plain
596 mm
206 kg
Light Gray
616 mm
Swing, Right-hinged
3
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers