missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Safety Storage Cabinet Q-CLASSIC-30 with 2 Doors
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3100306133647
Additional Details : Weight : 281.63879kg
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
1626 mm | |
4 x drawer (sheet steel powder-coated) | |
Safe and approved storage of flammable hazardous substances in working areas | |
519 mm | |
Steel | |
620 x 1164 x 1947 mm | |
519 x 1050 x 1626 mm | |
620 mm | |
Wing | |
90 mm | |
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
Body: Antharcite grey, Door: Light grey RAL 7035; Sheet Steel RAL 7035 | |
Safety storage cabinet | |
1050 mm | |
Powder-coated textured | |
1164 mm | |
302 kg | |
Anthracite Gray, Light Gray | |
1947 mm | |
Q30.195.116 | |
2 |