missing translation for 'onlineSavingsMsg'
Learn More

Biochrom™ Type 0 UV Visible Standard Cell

Brand:  Biochrom™ 80-2109-80

Code : GX

Additional Details : Weight : 0.14000kg

 View more versions of this product

Product Code. 11735195

  • £975.00 / Pack of 8


Call for latest availability
+44 (0)1509 555 500
Add to basket

If this item is out of stock, delivery will be in 3-4 working days providing the item is in stock at the supplier.
Please enquire with customer services on 01509 555500 for stock availability.

 

Description

Description

Standard rectangular cell with lid for visible/UV spectrophotometry. Minimum working volume 2mL, 10mm pathlength, 10mm internal width, 12. 5mm × 45mm external dimensions.
Specifications

Specifications

2mL
8Pack
Rectangular
Type 0 UV Visible Standard Cell
Visible/UV spectrophotometry
10mm
10mm
Videos