Learn More
Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration
Formulated for resistance to flex fatigue and abrasion and is less permeable than rubber tubing. Saint Gobain Life Sciences Tygon™ LMT-55 Tubing with Three Stop Configuration handles virtually all inorganic chemicals found in the lab.
Brand: Saint Gobain Life Sciences 070535-08-ND
Code : 02
Additional Details : Weight : 0.01000kg
Description
- Bio-based formulation delivers excellent performance
- Lot-to-lot consistency for reproducible results
- Non-toxic and non-contaminating
- Smooth, polished inner wall
- Slips easily over fittings and grips securely for simple lab set-ups
- Contains no BPA or phthalates
- Standard sizes available to hold full vacuum at room temperature
- General laboratory
- Food & beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Ideal for condensers, incubators, desiccators, gas lines and drain line
Specifications
Black/Black | |
Tygon R3607 | |
0.85 mm | |
-50°C | |
0.76 x 0.85mm | |
74°C |
0.76 mm | |
38.1 cm | |
For sample and reagent dispensing, biological and chemical assays, and water and soil analysis | |
12 Pack | |
Black/Black |