missing translation for 'onlineSavingsMsg'
Learn More

Heidolph instruments Tygon™ (Hydrocarbon) Tubing

Especially suited for hydrocarbons, mineral oil products and distillates. Heidolph™ Tygon™ Hydrocarbon Tubing is the ideal choice for petroleum products and more.

Brand:  Heidolph instruments 525-70027-00

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 10708992

  • £37.00 / Each
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

  • Ideal for petrol, kerosene, heating oil, cutting solutions and coolants with a glycol base.
  • Resistant to ozone and UV.
Specifications

Specifications

Autoclavable
Borosilicate Glass
1.6 mm
Non-sterile
4.8 mm
8 mm
Ideal for Petrol, Kerosene, Heating Oil, Cutting Solutions and Coolants with a Glycol Base.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers