Learn More
Masterflex™ Tygon™ 3 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 3 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
Brand:  Masterflex™ 95605-46
Code : A0
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 11927965
  • £79.65 / Pack of 12
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Specifications
Purple/Purple/Orange
Tygon F4040
12 Pack
2.54 mm
0.85 mm