Learn More
Masterflex™ Tygon™ 3 Stop Hydrocarbon Tubing
Tailored for petro-chemicals and resists swelling and cracking. Tygon™ 3 Stop Hydrocarbon Tubing is also UV and Ozone resistant.
Brand:  Masterflex™ 95605-40
Code : A0
Additional Details : Weight : 0.04000kg
 View more versions of this product
Product Code. 11997955
  • £80.90 / Pack of 12
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
Description
Description
  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easy to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant
Specifications
Specifications
Green/Green/Green
Tygon F4040
12 Pack
1.85 mm
0.85 mm