missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Performance Plastics™ Tygon™ F-4040-A Tubing

Easily identifiable yellow, flexible tubing for transportation of petroleum, heating oils, kerosene and glycol based coolants. Tygon™ F4040-A Tubing resists swelling and brittleness. UV and Ozone resistant.

Brand:  Saint Gobain Performance Plastics™ T401423

Code : 02

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11968099

  • £178.57 / 15m


Call for latest availability
+44 (0)1509 555 500
Add to basket

Alert:

Not for use with strong acids or alkalis.


Description

Description

Features and Benefits

  • Resists embrittlement
  • Compatible with most hydrocarbons
  • Resistant to swelling
  • Highly flexible, easty to install
  • Specifically developed for fuels and lubricants
  • Ozone and UV light resistant

Typical Applications

  • Small engine fuel lines
  • General automotive
  • Recreational vehicles
  • Lawn and garden equipment
  • Coolant transfer
  • Heating fuels
  • Cutting compounds
  • Polishing equipment
  • Lubrication lines
Specifications

Specifications

EU 2002/95/EC (ROHS) standard
11.1mm
14.3mm
1.6mm
Flexible hose designed to transfer fuel, lubricant and other petroleum-based Products
1.03 bar at 23°C
–37°C
178mm Hg at 23°C
Translucent Yellow
Tygon
15m
Flexible
57° Shore (A)
74°C
1 Pack
Videos