missing translation for 'onlineSavingsMsg'
Learn More

Ismatec™ Tygon™ E-LFL Microbore 2-Stop Tube Sets

Longest life of all Tygon™ formulations

Brand:  Ismatec™ 06447-12

Code : NEW

Additional Details : Weight : 0.04000kg

 View more versions of this product

Product Code. 15540652

  • £85.03 / Pack of 12


Estimated Shipment: 08-10-2021
Añadir a la cesta

Descripción

Descripción

  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
  • For use with Masterflex™ L/S™ multichannel pump heads
Especificaciones

Especificaciones

USP; EU Food 10/2011; European Pharmacopoeia; Non-DEHP; Non-Phthalate; RoHS; Prop 65 NA (no Prop 65 chemicals)
25.3 psi
Tygon E-LFL
0.38 m
0°C
12
40°C
0.19 mm
1.25 ft.
L/S Series Stopped Pump Tubing
Pk. of 12
1.65 mL/min
Videos
SDS
Documentos
Certificados