Learn More
Tocris Bioscience™ TY 52156
S1P3 receptor antagonist
POA
Specifications
Product Type | TY 52156 |
---|---|
Description | TY 52156 |
Molecular Weight (g/mol) | 364.27 |
Description
S1P3 receptor antagonist (Ki = 110 nM); inhibits S1P-induced Ca2+ release in HUVEC cells. Suppresses FTY-720-induced S1P3 receptor-mediated bradycardia in vivo. Also inhibits S1P-induced breast cancer stem cell expansion in vitro. Orally bioavailable.Specifications
TY 52156 | |
TY 52156 | |
364.27 |
For Research Use Only