missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TXK Recombinant Human Protein
TXK is a close relative of BMS, ITK and BTK.
Brand: Invitrogen™ PV5861
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TXK signals downstream of antigen receptors and other types of receptors.Specifications
NP_003319.2 | |
58.5kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Full Length | |
TEC Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
1mg | |
RUO - research use only | |
TXK | |
Human | |
GST Tag | |
Frozen |