missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, Acros Organics™

Brand:  Acros Organics 233360051

Code : 29

Additional Details : CAS Number : 9005-64-5 Weight : 5.55000kg

Quantity 5L
Packaging Plastic Bottle

 View more versions of this product

Product Code. 10503874

  • £116.20 / 5 L


EU Stock 52
Add to basket

Specifications

Chemical Identifiers

Polysorbate 20
9005-64-5
MFCD00165986
Polyoxyethylene(20)sorbitan monolaurate

Specifications

Polysorbate 20
9005-64-5
10ppm max.
96 to 108mg KOH/g
MFCD00165986
Polyoxyethylene(20)sorbitan monolaurate
1.4685 to 1.4715
Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
3% Max.
Clear colorless to pale yellow
5L
2mg KOH/g max.
1.1000g/mL
Liquid
Authentic
Plastic Bottle
Solubility in water: 100g/L. Other solubilities: 100g/L isopropylalcohol, xylene, insoluble in mineral oil, 10g/L propylene glycol
40 to 50mg KOH/g
400 mPa.s (25°C)
> 100.0°C
>150°C
1.1
Videos
SDS