Learn More
Thermo Scientific™ TurboFect Transfection Reagent
Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.
Brand:  Thermo Scientific™ R0541
UNSPSC : 41116133
Code : 88
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10314780
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Description
Highlights
  • High transfection efficiency of plasmid DNA into a wide variety of cell types
    • Adherent and suspension cells
    • Differentiated and undifferentiated cells
  • Excellent transfection efficiency in the presence or absence of serum 
  • Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
  • Simple protocol
Review our experimental considerations to improve your plasmid transfection results.
Review our experimental considerations to improve your plasmid transfection results.
Description & Specifications
Specifications
DNA transfection.siRNA-DNA co-transfection.
120µL
Alternative Products