Learn More
Thermo Scientific™ TurboFect™ Cell Transfection Reagent
Deliver DNA plasmids and expression vectors into eukaryotic cells with this highly efficient, easy-to-use, non-immunogenic transfection reagent.
Brand:  Thermo Scientific™ R0532
Code : PN
Additional Details : Weight : 1.81440kg
 View more versions of this product
Product Code. 12311863
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Description
Description
TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. The polymer forms compact, stable, positively charged complexes with DNA. These complexes protect DNA from degradation and facilitate gene delivery into eukaryotic cells. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells. Transfection can be performed in the presence or absence of serum. TurboFect Reagent demonstrates superior transfection efficiency and minimal toxicity when compared to other lipid-based or polymer-based transfection reagents.
Highlights:
  • High transfection efficiency of a wide variety of cell types
  • No optimization required for most cell lines
  • Excellent transfection efficiency in the presence or absence of serum
  • Minimal cytotoxicity
  • Ready-to-use – no need to reconstitute, dilute or manipulate
Recommended for:
Stable and transient transfection or co-transfection of DNA or RNA; Experiments with adherent or suspension cells, differentiated or undifferentiated cells; Gene expression studies and assays involving GFP or luciferase reporters
Specifications
Specifications
TurboFect Transfection Reagent
5 x 1mL
2500 transfections (25 standard 96-well plates)
TurboFect™ Transfection Reagent
Proprietary cationic polymer in water
TurboFect™ Transfection Reagent
Transfection
Plasmid DNA
Videos