missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten zirconium oxide, 99.7% (metals basis excluding Hf), Hf <0.38%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Tungsten zirconium oxide is used as a super acid catalyst in the hydrocarbon isomerization process and esterification of waste acid oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Green to White |
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Formula Weight | 586.92 |
Percent Purity | 99.7% |
Odor | Odorless |
Density | 5.10 g/mL |
Assay | (metals basis excluding Hf), Hf <0.3% |
Chemical Name or Material | Tungsten zirconium oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'