missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tungsten wire, gold plated, 0.1mm (0.004 in.) dia., W 99.95% (metals basis), Au 99.99%

Brand:  Alfa Aesar™ 045086.G1

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 1m

 View more versions of this product

Product Code. 15493935

  • £84.80 / 1m


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

Tungsten wire, gold plated
0.1mm (0.004 in.) dia.
MFCD00011461

Specifications

Tungsten wire, gold plated
1m
Au 99.99%, W 99.95%
99.95%
MFCD00011461
≈0.15g/m (W only), 3 to 5 wt% Au plating
0.1mm (0.004 in.) dia.
Wire
(metals basis)
Videos
SDS