Learn More
Tungsten wire, gold plated, 0.05mm (0.002in) dia, W 99.95% (metals basis), Au 99.99%, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041039.G1
48.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tungsten wire, gold plated | |
(metals basis) | |
1 m | |
Au 99.99% | |
Wire |
0.05mm (0.002 in.) dia. | |
MFCD00011461 | |
99.95% | |
≈0.02g/m (W only), 3 to 5 wt% Au plating |
RUO – Research Use Only