missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tungsten(VI) isopropoxide, 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles

Brand:  Alfa Aesar™ 041843.AE

Code : NEW

Additional Details : CAS Number : 52321-90-1 Weight : 0.01000kg

Quantity 100mL

 View more versions of this product

Product Code. 11314817

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Tungsten(VI) isopropoxide
W[OCH(CH3)2]6
MFCD00210652
52321-90-1
538.38
UN1219

Specifications

Tungsten(VI) isopropoxide
Liquid
ChemSeal™ bottles
MFCD00210652
5% w/v in isopropanol
Moisture Sensitive
52321-90-1
W[OCH(CH3)2]6
538.38
UN1219
100mL
Documentation

Technical Documents