missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Yellow |
Flash Point | 17°C (63°F) |
Odor | Alcohol-like |
Linear Formula | W(OC2H5)6 |
Quantity | 100 mL |
UN Number | UN1170 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethyl acetate,hydrocarbons and esters. |
Formula Weight | 454.21 |
Percent Purity | 99.8% |
Show More |
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'