missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Tungsten Titanium, 99.9% (metals basis)

Brand:  Alfa Aesar™ 036304.22

Code : NEW

Additional Details : CAS Number : 58397-70-9 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11313388

  • £412.34 / 100g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Tungsten Titanium
58397-70-9
MFCD02091752

Specifications

W:Ti; 90:10 wt%
100g
Sintered Lump
(metals basis)
3 to 12mm
Tungsten Titanium
58397-70-9
99.9%
MFCD02091752
Videos
SDS