Learn More
Thermo Scientific Alfa Aesar Tungsten rod, 4.0mm (0.16 in.) dia., Puratronic™, 99.96% (metals basis)
Brand: Thermo Scientific Alfa Aesar 011446.HR
Tungsten rod, 4.0mm (0.16 in.) dia., Puratronic™, 99.96% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-33-7 Weight : 0.01000kg