missing translation for 'onlineSavingsMsg'
Learn More

Tungsten plate, 6.4mm (0.25in) thick, 25x100mm (0.98x3.9in), 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 15404735
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 7440-33-7
Molecular Formula W
MDL Number MFCD00011461
Quantity 1 Pc.
Synonym wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES [W]
IUPAC Name tungsten
Molecular Weight (g/mol) 183.84
PubChem CID 23964
ChEBI CHEBI:27998
Percent Purity 99.95%
Weight ≈310g/plate
Form Plate
Assay (metals basis)
Chemical Name or Material Tungsten plate, 6.4mm (0.25 in.) thick, 25x100mm (0.98x3.9 in.)
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'