missing translation for 'onlineSavingsMsg'
Learn More

Tungsten gauze, 30 mesh woven from 0.1018mm (0.004in) dia wire, Thermo Scientific Chemicals

Catalog Number 11305227
87.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 75 x 75 mm
Mesh Size 30 Mesh
Form Wire Cloth
Chemical Name or Material Tungsten gauze, Woven from 0.1018mm (0.004 in.) dia. wire

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'