missing translation for 'onlineSavingsMsg'
Learn More

Tungsten gauze, 100 mesh woven from 0.0254mm (0.001in) dia wire, Thermo Scientific Chemicals

Catalog Number 11325227
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
This item is not returnable. View return policy
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]
This item is not returnable. View return policy
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsten
[W]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 75 x 75 mm
Mesh Size 100 Mesh
Form Wire Cloth
Chemical Name or Material Tungsten gauze, Woven from 0.0254mm (0.001 in.) dia. wire

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'