Learn More
RETSCH Tungsten Carbide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Tungsten Carbide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand:  RETSCH 01.462.0155
Code : 50
Additional Details : Weight : 2.70000kg
 View more versions of this product
Product Code. 10791413
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Available in a variety of sizes
  • Optional aeration covers
  • Pressure tight safety closure devices
Specifications
Specifications
Comfort Grinding Jar
125 mL
For PM 100, PM 200, PM 400 and S 100
Tungsten Carbide
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products