Learn More
Retsch™ Tungsten Carbide Comfort Grinding Jars
Use these grinding jars with Retsch™ ball mills. Retsch™ Tungsten Carbide Comfort Grinding Jars feature an O-type sealing for safe operation.
Brand:  Retsch™ 01.462.0156
Code : 50
Additional Details : Weight : 1.85000kg
 View more versions of this product
Product Code. 10619881
  • £3166.67 / Each
Estimated Shipment
27-02-2018
Add to basket
Specifications
Specifications
50mL
Tungsten Carbide
For PM 100, PM 200, PM 400 and S 100
Comfort Grinding Jar
Videos
View accessory products