Learn More
Contec™ TuffStuff™ Critical Task Wipes
Maintain strength even when wet. Contec™ TuffStuff™ Critical Task Wipes are highly absorbent, low-linting, and very durable.
Brand: Contec™ TSWC1214
Code : NEW
Additional Details : Weight : 2.00000kg
Description
- Ideal for a variety of tough jobs including general industrial cleaning, production line and heavy equipment maintenance, and hand and tool cleanup
- Constructed of cellulose and polyester
- Can be used with harsh solvents such as isopropyl alcohol, MEK, and MPK without falling apart
- Contain no binders, chemical residue, or contaminants that can result in surface damage or rework
- Recommended for critical cleaning in biotech and pharmaceutical industries
Specifications
Wipes | |
ISO Class 6-8 environments | |
500/Roll, 1 Roll/Dispenser |
Polyester Cellulose | |
30.5 x 33 cm | |
Non-sterile |