missing translation for 'onlineSavingsMsg'
Learn More

beta-3 Tubulin, clone: 2G10-TB3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-4510-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15527446

  • £57.93 / 25µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 2G10 monoclonal antibody recognizes beta tubulin class III, a member of the large family of tubulins that are components of the microtubules of the cytoskeletal network. Beta tubulin class III expression is exclusively found in neuronal cells of adult mammals. Although expression has been mainly found in neuronal cells, during fetal development, transient expression has been documented in both glial and neuronal precursors localized to the subventricular zone. Identification of beta tubulin class III expression is useful as a structural marker for developing and mature neural cells. Given the high degree of conservation of this protein across species, the 2G10 clone is expected to recognize human, mouse, rat, and bovine beta tubulin class III.Applications Reported: This 2G10 antibody has been reported for use in immunoprecipitation, western blotting, immunohistochemical staining of frozen or formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.Applications Tested: This 2G10 antibody has been tested by immunocytochemistry of fixed and permeabilized cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Tubulin
2G10-TB3
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TUBB3
IgG2a
RUO
Primary
Rat
Videos