Learn More
beta-3 Tubulin Monoclonal Antibody (2G10-TB3), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 53-4510-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The 2G10 monoclonal antibody recognizes beta tubulin class III, a member of the large family of tubulins that are components of the microtubules of the cytoskeletal network. Beta tubulin class III expression is exclusively found in neuronal cells of adult mammals. Although expression has been mainly found in neuronal cells, during fetal development, transient expression has been documented in both glial and neuronal precursors localized to the subventricular zone. Identification of beta tubulin class III expression is useful as a structural marker for developing and mature neural cells. Given the high degree of conservation of this protein across species, the 2G10 clone is expected to recognize human, mouse, rat, and bovine beta tubulin class III. Applications Reported: This 2G10-TB3 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. Applications Tested: This 2G10-TB3 antibody has been tested by immunocytochemistry of formaldehyde-fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
beta-3 Tubulin Monoclonal antibody specifically detects beta-3 Tubulin in Bovine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
beta-3 Tubulin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q13509, Q2T9S0, Q4QRB4, Q9ERD7 | |
TUBB3 | |
Affinity chromatography | |
RUO | |
10381, 22152, 246118, 768070 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
2G10-TB3 | |
Alexa Fluor 488 | |
TUBB3 | |
3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
Mouse | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
For Research Use Only.