Learn More
Masterflex™ Straight Adapter, Barb x NPT(M), HDPE
Cole-Parmer™ Straight Adapter, Barb x NPT(M), HDPE adapts from an NPT male thread to a hose barb and provides a quick, easy, and reliably secure connection.
Brand: Masterflex™ 06444-18
Code : XX
Additional Details : Weight : 0.01000kg
Description
- Black, HDPE construction
- Easy to use
- Sterilizable via autoclave or ETO
Specifications
NPT Male Adapter to Hose Barb, 1/4 in. NPT to 3/16 in. ID Tubing | |
HDPE | |
10 Pack | |
Fits tubing that accepts 1/4 in. NPT connection, 3/16 in. ID tubing | |
14 mm |
Autoclavable | |
1/4 in. NPT connection, 3/16 in. ID tubing | |
Barb x NPT(M) Connector Fittings | |
Autoclave or Ethylene Oxide | |
Black |