missing translation for 'onlineSavingsMsg'
Learn More

TTP 22, Tocris Bioscience™

High affinity, selective CK2 inhibitor

Brand:  RD Systems (Bio-Techne) 4432/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464744

  • £165.00 / 10mg


Estimated Shipment
03-01-2019
Add to basket

Description

Description

High affinity, ATP-competitive casein kinase 2 (CK2) inhibitor (IC50 = 0.1μM, Ki = 40 nM). Displays selectivity for CK2 over JNK3, ROCK1 and MET (no inhibitory effects at 10μM).
Specifications

Chemical Identifiers

329907-28-0
330.42
ttp 22, ttp22, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanylpropanoic acid, ttp-22, 3-5-p-tolyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanyl propanoic acid, 7ey, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thiopheno 3,2-e pyrimidin-4-ylthio propanoic acid
3-[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanylpropanoic acid
C16H14N2O2S2
RAOULLCLLOGTDA-UHFFFAOYSA-N
1536915
CC1=CC=C(C=C1)C2=CSC3=C2C(=NC=N3)SCCC(=O)O

Specifications

TTP 22
329907-28-0
ttp 22, ttp22, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanylpropanoic acid, ttp-22, 3-5-p-tolyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl sulfanyl propanoic acid, 7ey, 3-5-4-methylphenyl thieno 2,3-d pyrimidin-4-yl thio propanoic acid, 3-5-4-methylphenyl thiopheno 3,2-e pyrimidin-4-ylthio propanoic acid
CC1=CC=C(C=C1)C2=CSC3=C2C(=NC=N3)SCCC(=O)O
330.42
330.42
10mg
C16H14N2O2S2
RAOULLCLLOGTDA-UHFFFAOYSA-N
3-[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanylpropanoic acid
1536915
>98%
Videos