missing translation for 'onlineSavingsMsg'
Learn More

TSLP Receptor Mouse anti-Human, PerCP-eFluor™ 710, Clone: eBio1A6 (1A6), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5499-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15599266

  • £241.00 / 100 tests

Estimated Shipment
07-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor® 710 can be used in place of PE-Cy5, PE-Cy5.5 or PerCP-Cy5.5. PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm). Please make sure that your instrument is capable of detecting this fluorochrome. For a filter configuration, we recommend using the 685 LP dichroic mirror and 710/40 band pass filter, however the 695/40 band pass filter is an acceptable alternative. Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintece of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.
Specifications

Specifications

TSLP Receptor
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9HC73
WI2-87105E3.1, CRL2, CRLF2Y, TSLPR; IL-XR; P2RY8/CRLF2 fusion; TSLP receptor; cytokine receptor CRL2 precusor; cytokine receptor-like 2; thymic stromal lymphopoietin protein receptor; thymic stromal lymphopoietin receptor; thymic stromal-derived lymphopoietin receptor
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBio1A6 (1A6)
PerCP-eFluor™ 710
CRLF2
Liquid
CRLF2
IgG2a, lambda
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
64109
Videos
SDS
Documents

For Research Use Only.