missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TSG Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | TSG |
---|---|
Clone | 120024 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
TSG Monoclonal specifically detects TSG in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TSG | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
TSGtwisted gastrulation protein homolog 1, twisted gastrulation homolog 1 (Drosophila), TWSG1 | |
TWSG1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
120024 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rat | |
Mouse | |
Q9EP52 | |
57045 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TSG Cys25-Phe222 Accession # Q9EP52 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse TSG in direct ELISAs and Western blots. |
For Research Use Only