missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse/Rat TSC22 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | TSC22/TSC22D1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
TSC22/TSC22D1 Polyclonal specifically detects TSC22/TSC22D1 in Mouse, Rat samples. It is validated for Western Blot.Specifications
TSC22/TSC22D1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse, Rat | |
Cerebral protein 2, Egr5, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1, TSC22DKFZp686O19206 | |
TSC22D1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P62500.2 | |
8848 | |
E. coli-derived recombinant mouse TSC22 Met1-Ala143 Accession # P62500.2 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse and rat TSC22 in direct ELISAs and Western blots. |
For Research Use Only