missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophol, 97%, Thermo Scientific Chemicals
11.53 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a reactant for preparation of inhibitors of the C-terminal domain of RNA polymerase II and their antitumor activities, anti-HIV-1 agents, inhibitors of Protein-Protein Interactions, partial agonists of the serotonin 5-HT1A receptor, growth hormone secretagogues, vascular endothelial growth factor (VEGF) inhibitors, A2B adenosine receptor ligands, potential detoxification inhibitors of the crucifer phytoalexin brassinin, inhibitors of interleukin and dual binding site acetyl cholinesterase inhibitors.
Solubility
Solubility in methanol gives very faint turbidity. Soluble in water at 25°C 2749 mg/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is employed as a reactant for preparation of inhibitors of the C-terminal domain of RNA polymerase II and their antitumor activities, anti-HIV-1 agents, inhibitors of Protein-Protein Interactions, partial agonists of the serotonin 5-HT1A receptor, growth hormone secretagogues, vascular endothelial growth factor (VEGF) inhibitors, A2B adenosine receptor ligands, potential detoxification inhibitors of the crucifer phytoalexin brassinin, inhibitors of interleukin and dual binding site acetyl cholinesterase inhibitors.
Solubility
Solubility in methanol gives very faint turbidity. Soluble in water at 25°C 2749 mg/L.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 56°C to 59°C |
Boiling Point | 174°C (2 mmHg) |
Flash Point | 255°C (491°F) |
Quantity | 1 g |
Beilstein | 125553 |
Merck Index | 14,9798 |
Formula Weight | 161.2 |
Percent Purity | 97% |
Chemical Name or Material | Tryptophol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'