missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan Hydroxylase 1/TPH-1 Antibody (007), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90021AF647
This item is not returnable.
View return policy
Description
Tryptophan Hydroxylase 1/TPH-1 Monoclonal specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for ELISA.
Specifications
| Tryptophan Hydroxylase 1/TPH-1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 7 | |
| ELISA | |
| EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Tryptophan Hydroxylase 1/TPH-1 (P17752-1; Ile2-Ile444). | |
| 0.1 mL | |
| Lipid and Metabolism, Neuroscience, Neurotransmission, Phospho Specific | |
| 7166 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction