missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Tryptase gamma-1/TPSG1 Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | Tryptase gamma-1/TPSG1 |
---|---|
Clone | 279005 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Tryptase gamma-1/TPSG1 Monoclonal specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Tryptase gamma-1/TPSG1 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rat | |
Human | |
Q9NRR2 | |
25823 | |
Mouse myeloma cell line NS0-derived recombinant human Tryptase gamma-1/TPSG1 Arg20-Arg281 Accession # Q9NRR2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Tryptase gamma-1/TPSG1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MCPT-1, recombinant human (rh) Tryptase alpha , rmTryptase beta-1, rhTryptase beta-2, or rmTryptase-5 is observed. |
279005 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, TPSG1, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II, Tryptase gamma1 | |
TPSG1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only