Alfa Aesar™ Trypsin, porcine pancreas

A pancreatic serine protease

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J60402.09

UNSPSC: 12352204

Code: NEW

Additional Details:
Additional Details: CAS Number: 9002-07-7 Weight: 0.01000kg



Product Code. 15433179

Quantity Price
1 £ 43.57 / 10g
Estimated Shipment date
from Supplier 05-05-2017
Add to basket


Description and Specification

Specification

Format Lyophilized Powder
Description Off-white to Pale Brown
For Use With (Application) A pancreatic serine protease
Product Type Trypsin
Molecular Weight 23.8 kd
Quantity 10g
Storage Requirements −30° to −10°C
CAS 9002-07-7
Synonym EC 3.4.21.4; Trypsin IV porcine

Reconstitute in aqueous buffer

Documentation

Technical documents