missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trypsin, porcine pancreas

A pancreatic serine protease

Brand:  Alfa Aesar™ J60402.18

UNSPSC : 12352204

Code : NEW

Additional Details : CAS Number : 9002-07-7 Weight : 0.05000kg

 View more versions of this product

Product Code. 15443179

  • £153.59 / 50g


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications

Description

Reconstitute in aqueous buffer
Description & Specifications

Specifications

Lyophilized Powder
A pancreatic serine protease
23.8 kd
−30° to −10°C
EC 3.4.21.4; Trypsin IV porcine
Off-white to Pale Brown
Trypsin
50g
9002-07-7
Documentation

Technical Documents